Anti-PIBF1 Mouse Monoclonal Antibody [clone: 7A3]

Supplier: Novus Biologicals
H00010464-M01
103338-958EA 798.46 USD
103338-958
Anti-PIBF1 Mouse Monoclonal Antibody [clone: 7A3]
Antibodies
The PIBF1 Antibody (7A3) from Novus Biologicals is a mouse monoclonal antibody to PIBF1. This antibody reacts with human. The PIBF1 Antibody (7A3) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PIBF1
Clonality: Monoclonal
Clone: 7A3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR