Anti-PIAS1 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00008554-B01P
103335-166EA 399 USD
103335-166
Anti-PIAS1 Mouse Polyclonal Antibody
Antibodies
The PIAS1 Antibody from Novus Biologicals is a mouse polyclonal antibody to PIAS1. This antibody reacts with human. The PIAS1 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: PIAS1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR