Anti-PI4KB/PI4KIII beta Mouse Monoclonal Antibody [clone: 3B1]

Supplier: Novus Biologicals
H00005298-M02
103307-490EA 798.46 USD
103307-490
Anti-PI4KB/PI4KIII beta Mouse Monoclonal Antibody [clone: 3B1]
Antibodies
The PI4KB / PI4KIII beta Antibody (3B1) from Novus Biologicals is a mouse monoclonal antibody to PI4KB / PI4KIII beta. This antibody reacts with human. The PI4KB / PI4KIII beta Antibody (3B1) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PI4KB/PI4KIII beta
Clonality: Monoclonal
Clone: 3B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR