Anti-PI16 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-19772
102710-416EA 635.51 USD
102710-416
Anti-PI16 Rabbit Polyclonal Antibody
Antibodies
The Peptidase Inhibitor 16 / PI16 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Peptidase Inhibitor 16 / PI16. This antibody reacts with human. The Peptidase Inhibitor 16 / PI16 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: PI16
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR