Anti-PHYHIPL Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-84858
103260-756EA 734.58 USD
103260-756
Anti-PHYHIPL Rabbit Polyclonal Antibody
Antibodies
The PHYHIPL Antibody from Novus Biologicals is a rabbit polyclonal antibody to PHYHIPL. This antibody reacts with human. The PHYHIPL Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PHYHIPL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR