Anti-PHYH Mouse Monoclonal Antibody [clone: 1F2-5B9]

Supplier: Novus Biologicals
H00005264-M01
103307-394EA 798.46 USD
103307-394
Anti-PHYH Mouse Monoclonal Antibody [clone: 1F2-5B9]
Antibodies
The PHYH Antibody (1F2-5B9) from Novus Biologicals is a mouse monoclonal antibody to PHYH. This antibody reacts with human. The PHYH Antibody (1F2-5B9) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PHYH
Clonality: Monoclonal
Clone: 1F2-5B9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR