Anti-PHTF1 Mouse Monoclonal Antibody [clone: 3B7]

Supplier: Novus Biologicals
H00010745-M01
103339-518EA 798.46 USD
103339-518
Anti-PHTF1 Mouse Monoclonal Antibody [clone: 3B7]
Antibodies
The PHTF1 Antibody (3B7) from Novus Biologicals is a mouse monoclonal antibody to PHTF1. This antibody reacts with human. The PHTF1 Antibody (3B7) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PHTF1
Clonality: Monoclonal
Clone: 3B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR