Anti-PHOX2A Mouse Monoclonal Antibody [clone: 4F6]

Supplier: Novus Biologicals
H00000401-M01
103322-146EA 798.46 USD
103322-146
Anti-PHOX2A Mouse Monoclonal Antibody [clone: 4F6]
Antibodies
The PHOX2A Antibody (4F6) from Novus Biologicals is a mouse monoclonal antibody to PHOX2A. This antibody reacts with human, rat. The PHOX2A Antibody (4F6) has been validated for the following applications: Western Blot, ELISA, Immunocytochemistry / Immunofluorescence, RNA Inhibition.

Type: Primary
Antigen: PHOX2A
Clonality: Monoclonal
Clone: 4F6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR