Anti-PHKG1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-87280
103270-562EA 734.58 USD
103270-562
Anti-PHKG1 Rabbit Polyclonal Antibody
Antibodies
The PHKG1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PHKG1. This antibody reacts with human, mouse, rat. The PHKG1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PHKG1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR