Anti-PHKG1 Mouse Monoclonal Antibody [clone: 2G6]

Supplier: Novus Biologicals
H00005260-M01
103307-336EA 798.46 USD
103307-336
Anti-PHKG1 Mouse Monoclonal Antibody [clone: 2G6]
Antibodies
The PHKG1 Antibody (2G6) from Novus Biologicals is a mouse monoclonal antibody to PHKG1. This antibody reacts with human. The PHKG1 Antibody (2G6) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PHKG1
Clonality: Monoclonal
Clone: 2G6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR