Anti-PHGDH Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00026227-B01P
103342-190EA 399 USD
103342-190
Anti-PHGDH Mouse Polyclonal Antibody
Antibodies
The PHGDH Antibody from Novus Biologicals is a mouse polyclonal antibody to PHGDH. This antibody reacts with human, mouse. The PHGDH Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PHGDH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR