Anti-PHF1 Mouse Monoclonal Antibody [clone: 2A12]

Supplier: Novus Biologicals
H00005252-M03
103332-728EA 798.46 USD
103332-728
Anti-PHF1 Mouse Monoclonal Antibody [clone: 2A12]
Antibodies
The PHF1 Antibody (2A12) from Novus Biologicals is a mouse monoclonal antibody to PHF1. This antibody reacts with human. The PHF1 Antibody (2A12) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PHF1
Clonality: Monoclonal
Clone: 2A12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR