Anti-PHD4 Rabbit Polyclonal Antibody (Alexa Fluor® 700)

Supplier: Novus Biologicals
NB100-295AF700
103357-662EA 706.28 USD
103357-662
Anti-PHD4 Rabbit Polyclonal Antibody (Alexa Fluor® 700)
Antibodies
The PHD4 / HIF Prolyl Hydroxylase 4 Antibody [Alexa Fluor« 700] from Novus Biologicals is a rabbit polyclonal antibody to PHD4 / HIF Prolyl Hydroxylase 4. This antibody reacts with human, rat, mouse (negative). The PHD4 / HIF Prolyl Hydroxylase 4 Antibody [Alexa Fluor« 700] has been validated for the following applications: Western Blot, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PHD4
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 700
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat, Mouse (Negative)
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR