Anti-PGM3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00005238-D01P
103332-696EA 429 USD
103332-696
Anti-PGM3 Rabbit Polyclonal Antibody
Antibodies
The PGM3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PGM3. This antibody reacts with human. The PGM3 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: PGM3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR