Anti-PGM1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-85982
103269-166EA 734.58 USD
103269-166
Anti-PGM1 Rabbit Polyclonal Antibody
Antibodies
The PGM1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PGM1. This antibody reacts with human, mouse. The PGM1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PGM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR