Anti-PGD2 Mouse Polyclonal Antibody

H00005730-B01P
103308-616EA 583.08 USD
103308-616
Anti-PGD2 Mouse Polyclonal Antibody
The PGD2 Synthase / PTGDS Antibody from Novus Biologicals is a mouse polyclonal antibody to PGD2 Synthase / PTGDS. This antibody reacts with human. The PGD2 Synthase / PTGDS Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: PGD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR