Anti-PGD Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-85925
103272-276EA 734.58 USD
103272-276
Anti-PGD Rabbit Polyclonal Antibody
Antibodies
The PGD Antibody from Novus Biologicals is a rabbit polyclonal antibody to PGD. This antibody reacts with human, mouse. The PGD Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PGD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR