Anti-PGCP Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00010404-B01P
103338-812EA 798.46 USD
103338-812
Anti-PGCP Mouse Polyclonal Antibody
Antibodies
The PGCP Antibody from Novus Biologicals is a mouse polyclonal antibody to PGCP. This antibody reacts with human. The PGCP Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PGCP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR