Anti-PGC1A Mouse Monoclonal Antibody [clone: 3G11]

H00010891-M12
103339-746EA 583.08 USD
103339-746
Anti-PGC1A Mouse Monoclonal Antibody [clone: 3G11]
The PGC1 alpha Antibody (3G11) from Novus Biologicals is a mouse monoclonal antibody to PGC1 alpha. This antibody reacts with human. The PGC1 alpha Antibody (3G11) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PGC1A
Clonality: Monoclonal
Clone: 3G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR