Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PGC1A Mouse Monoclonal Antibody [clone: 4A8]

Supplier: Novus Biologicals
H00010891-M01
The PGC1 alpha Antibody (4A8) from Novus Biologicals is a mouse monoclonal antibody to PGC1 alpha. This antibody reacts with human. The PGC1 alpha Antibody (4A8) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PGC1A
Clonality: Monoclonal
Clone: 4A8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human




Log in to see your contract pricing and availability.
  Remember me on this device
 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 μg H00010891-M01 103339-734 Each (100µG) Retrieving Restricted

Temperature sensitive This Item is temperature sensitive and has specific temperature and storage requirements for shipping/delivery. Requires a maximum 2 day delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol PGC1A
Antigen Name PGC1 alpha
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone 4A8
Host Mouse
Isotype IgG2b Kappa
Western Blot Yes
ELISA Yes
Antigen Synonyms coactivator 1, Ligand effect modulator 6, gamma, PPAR gamma coactivator-1, PGC1A, PGC-1-alpha, LEM6, peroxisome proliferator-activated receptor gamma, coactivator 1 alpha, PPAR-gamma coactivator 1-alpha, PPARGC1PGC-1v, alpha, peroxisome proliferative activated receptor, PGC1peroxisome proliferative activated receptor, PPAR gamma coactivator variant form, PGC1APGC-1(alpha), ligand effect modulator-6, peroxisome proliferator-activated receptor gamma coactivator 1-alpha, PPARGC-1-alpha
Gene ID 10891
Immunogen PPARGC1A (NP_037393, 689 a.a. ~ 799 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.TRTELRDRFEVFGEIEECTVNLRDDGDSYGFITYRYTCDAFAALENGYTLRRSNETDFELYFCGRKQFFKSNYADLDSNSDDFDPASTKSKYDSLDFDSLLKEAQRSLRR
Purification IgG purified
Storage Buffer PBS (pH 7.4)
Storage Temperature Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty