Anti-PGBD2 Goat Polyclonal Antibody

Supplier: Novus Biologicals
NB100-93427
103408-980EA 564.74 USD
103408-980
Anti-PGBD2 Goat Polyclonal Antibody
Antibodies
The PGBD2 Antibody from Novus Biologicals is a goat polyclonal antibody to PGBD2. This antibody reacts with human. The PGBD2 Antibody has been validated for the following applications: Peptide ELISA.

Type: Primary
Antigen: PGBD2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-ESNADTTSQGRRSRR
Host: Goat
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR