Anti-PGA5 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00005222-D01P
103332-666EA 429 USD
103332-666
Anti-PGA5 Rabbit Polyclonal Antibody
Antibodies
The PGA5 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PGA5. This antibody reacts with human. The PGA5 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: PGA5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR