Anti-PFN2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
H00005217-D01P
103332-658EA 429 USD
103332-658
Anti-PFN2 Rabbit Polyclonal Antibody
Antibodies
The Profilin 2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Profilin 2. This antibody reacts with human. The Profilin 2 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: PFN2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR