Anti-PFN2 Mouse Monoclonal Antibody [clone: 5F11]

Supplier: Novus Biologicals
H00005217-M04
103332-660EA 798.46 USD
103332-660
Anti-PFN2 Mouse Monoclonal Antibody [clone: 5F11]
Antibodies
The Profilin 2 Antibody (5F11) from Novus Biologicals is a mouse monoclonal antibody to Profilin 2. This antibody reacts with human. The Profilin 2 Antibody (5F11) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PFN2
Clonality: Monoclonal
Clone: 5F11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR