Anti-PFN1 Rabbit Polyclonal Antibody

NBP2-19941
102710-754EA 493.85 USD
102710-754
Anti-PFN1 Rabbit Polyclonal Antibody
The Profilin 1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Profilin 1. This antibody reacts with human, mouse, rat. The Profilin 1 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PFN1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR