Anti-PFDN4 Mouse Monoclonal Antibody [clone: 2G4]

Supplier: Novus Biologicals
H00005203-M03
103332-624EA 798.46 USD
103332-624
Anti-PFDN4 Mouse Monoclonal Antibody [clone: 2G4]
Antibodies
The PFDN4 Antibody (2G4) from Novus Biologicals is a mouse monoclonal antibody to PFDN4. This antibody reacts with human. The PFDN4 Antibody (2G4) has been validated for the following applications: Western Blot, ELISA.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: PFDN4
Clonality: Monoclonal
Clone: 2G4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR