Anti-PEX19 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00005824-B01P
103308-822EA 399 USD
103308-822
Anti-PEX19 Mouse Polyclonal Antibody
Antibodies
The PEX19 Antibody from Novus Biologicals is a mouse polyclonal antibody to PEX19. This antibody reacts with human. The PEX19 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: PEX19
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR