Anti-PEX12 Goat Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-49893
103408-972EA 299 USD
103408-972
Anti-PEX12 Goat Polyclonal Antibody
Antibodies
The PEX12 Antibody from Novus Biologicals is a goat polyclonal antibody to PEX12. This antibody reacts with human, mouse, rat, bovine. The PEX12 Antibody has been validated for the following applications: Peptide ELISA.

Type: Primary
Antigen: PEX12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: RSHQACPITGYP - C Terminus
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR