Anti-PEX11A Mouse Monoclonal Antibody [clone: 3B1]

Supplier: Novus Biologicals
H00008800-M04
103335-640EA 798.46 USD
103335-640
Anti-PEX11A Mouse Monoclonal Antibody [clone: 3B1]
Antibodies
The PEX11A Antibody (3B1) from Novus Biologicals is a mouse monoclonal antibody to PEX11A. This antibody reacts with human. The PEX11A Antibody (3B1) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PEX11A
Clonality: Monoclonal
Clone: 3B1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR