Anti-PERQ2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-34129
103286-834EA 734.58 USD
103286-834
Anti-PERQ2 Rabbit Polyclonal Antibody
Antibodies
The PERQ2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PERQ2. This antibody reacts with human. The PERQ2 Antibody has been validated for the following applications: Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PERQ2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR