Anti-PEPD Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00005184-B01P
103332-590EA 399 USD
103332-590
Anti-PEPD Mouse Polyclonal Antibody
Antibodies
The PEPD Antibody from Novus Biologicals is a mouse polyclonal antibody to PEPD. This antibody reacts with human. The PEPD Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: PEPD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR