Anti-PECR Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-83198
103258-978EA 734.58 USD
103258-978
Anti-PECR Rabbit Polyclonal Antibody
Antibodies
The PECR Antibody from Novus Biologicals is a rabbit polyclonal antibody to PECR. This antibody reacts with human. The PECR Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PECR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR