Anti-PECI Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-19766
102710-404EA 635.51 USD
102710-404
Anti-PECI Rabbit Polyclonal Antibody
Antibodies
The PECI Antibody from Novus Biologicals is a rabbit polyclonal antibody to PECI. This antibody reacts with human. The PECI Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PECI
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR