Anti-PEAR1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-90449
103277-872EA 734.58 USD
103277-872
Anti-PEAR1 Rabbit Polyclonal Antibody
Antibodies
The PEAR1 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PEAR1. This antibody reacts with human. The PEAR1 Antibody has been validated for the following applications: Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.

Type: Primary
Antigen: PEAR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR