Anti-Pea3 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
20580002-0.1MG
103317-608EA 399 USD
103317-608
Anti-Pea3 Rabbit Polyclonal Antibody
Antibodies
The Pea3 Antibody from Novus Biologicals is a rabbit polyclonal antibody to Pea3. This antibody reacts with human. The Pea3 Antibody has been validated for the following applications: Western Blot, ELISA, Immunoprecipitation.

Type: Primary Antibodies
Antigen: Pea3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR