Anti-PDZRN4 Rabbit Polyclonal Antibody

NBP1-85144
103273-202EA 607.69 USD
103273-202
Anti-PDZRN4 Rabbit Polyclonal Antibody
The PDZRN4 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PDZRN4. This antibody reacts with human. The PDZRN4 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PDZRN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR