|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | PDXK |
Antigen Name | Pyridoxal Kinase |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Monoclonal |
Antibody Type | Primary |
Clone | 4G6 |
Host | Mouse |
Isotype | IgG2a Kappa |
Western Blot | Yes |
ELISA | Yes |
Antigen Synonyms | PNKhuman pyridoxal kinase, pyridoxamine kinase, C21orf97, EC 2.7.1.3510FLJ31940, pyridoxal kinase, Pyridoxine kinase, chromosome 21 open reading frame 124, EC 2.7.1.35, vitamin B6) kinase, FLJ21324, FLJ37311, DKFZp566A071, MGC15873, PKHchromosome 21 open reading frame 97, MGC52346, PRED79, C21orf124, MGC31754, pyridoxal (pyridoxine, vitamin B6 kinase |
Gene ID | 8566 |
Immunogen | PDXK (AAH00123, 1 a.a. - 312 a.a.) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MEEECRVLSIQSHVIRGYVGNRAATFPLQVLGFEIDAVNSVQFSNHTGYAHWKGQVLNSDELQELYEGLRLNNMNKYDYVLTGYTRDKSFLAMVVDIVQELKQQNPRLVYVCDPVLGDKWDGEGSMYVPEDLLPVYKEKVVPLADIITPNQFEAELLSGRKIHSQEEALRVMDMLHSMGPDTVVITSSDLPSPQGSNYLIVLGSQRRRNPAGSVVMERIRMDIRKVDAVFVGTGDLFAAMLLAWTHKHPNNLKVACEKTVSTLHHVLQRTIQCAKAQAGEGVRPSPMQLELRMVQSKRDIEDPEIVVQATVL |
Purification | Protein A purified |
Storage Buffer | PBS (pH 7.4) |
Storage Temperature | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |