Anti-PDPK2 Mouse Monoclonal Antibody [clone: 1G3]

Supplier: Novus Biologicals
H00653650-M01
103348-576EA 798.46 USD
103348-576
Anti-PDPK2 Mouse Monoclonal Antibody [clone: 1G3]
Antibodies
The PDPK2 Antibody (1G3) from Novus Biologicals is a mouse monoclonal antibody to PDPK2. This antibody reacts with human. The PDPK2 Antibody (1G3) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PDPK2
Clonality: Monoclonal
Clone: 1G3
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR