Anti-PDP1/PPAPDC2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-83443
103267-758EA 734.58 USD
103267-758
Anti-PDP1/PPAPDC2 Rabbit Polyclonal Antibody
Antibodies
The PDP1 / PPAPDC2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PDP1 / PPAPDC2. This antibody reacts with human. The PDP1 / PPAPDC2 Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunocytochemistry / Immunofluorescence, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PDP1/PPAPDC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR