Anti-PDP1 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP1-87309
103275-684EA 734.58 USD
103275-684
Anti-PDP1 Rabbit Polyclonal Antibody
Antibodies
The Pyruvate Dehydrogenase Phosphatase / PDP1 / PPM2C Antibody from Novus Biologicals is a rabbit polyclonal antibody to Pyruvate Dehydrogenase Phosphatase / PDP1 / PPM2C. This antibody reacts with human. The Pyruvate Dehydrogenase Phosphatase / PDP1 / PPM2C Antibody has been validated for the following applications: Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Type: Primary
Antigen: PDP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR