|
![]() |
ORDER
SPECIFICATIONS
Antigen Symbol | PDLIM7 |
Reactivity | Human |
Conjugation | Unconjugated |
Clonality | Polyclonal |
Antibody Type | Primary |
Host | Mouse |
Isotype | IgG |
Western Blot | Yes |
ELISA | Yes |
ImmunoFluorescence | Yes |
Antigen Synonyms | LIM domain protein, PDZ and LIM domain 7 (enigma), ENIGMAProtein enigma, LMP1, LMP, LIM mineralization protein, 1110003B01Rik, PDZ and LIM domain protein 7 |
Gene ID | 9260 |
Immunogen | PDLIM7 (NP_005442.2, 1 a.a. ~ 457 a.a) full-length human protein.MDSFKVVLEGPAPWGFRLQGGKDFNVPLSISRLTPGGKAAQAGVAVGDWVLSIDGENAGSLTHIEAQNKIRACGERLSLGLSRAQPVQSKPQKASAPAADPPRYTFAPSVSLNKTARPFGAPPPADSAPQQNGQPLRPLVPDASKQRLMENTEDWRPRPGTGQSRSFRILAHLTGTEFMQDPDEEHLKKSSQVPRTEAPAPASSTPQEPWPGPTAPSPTSRPPWAVDPAFAERYAPDKTSTVLTRHSQPATPTPL |
Purification | Protein A purified |
Storage Buffer | PBS (pH 7.4) |
Storage Temperature | Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |