Anti-PDLIM4 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00008572-B01P
103335-250EA 798.46 USD
103335-250
Anti-PDLIM4 Mouse Polyclonal Antibody
Antibodies
The PDLIM4 Antibody from Novus Biologicals is a mouse polyclonal antibody to PDLIM4. This antibody reacts with human. The PDLIM4 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: PDLIM4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR