Anti-PDLIM2 Rabbit Polyclonal Antibody

Supplier: Novus Biologicals
NBP2-19754
102710-380EA 635.51 USD
102710-380
Anti-PDLIM2 Rabbit Polyclonal Antibody
Antibodies
The PDLIM2 Antibody from Novus Biologicals is a rabbit polyclonal antibody to PDLIM2. This antibody reacts with human. The PDLIM2 Antibody has been validated for the following applications: Western Blot.

Type: Primary
Antigen: PDLIM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR