Anti-PDLIM1 Mouse Polyclonal Antibody

Supplier: Novus Biologicals
H00009124-B01P
103336-268EA 399 USD
103336-268
Anti-PDLIM1 Mouse Polyclonal Antibody
Antibodies
The PDLIM1 Antibody from Novus Biologicals is a mouse polyclonal antibody to PDLIM1. This antibody reacts with human. The PDLIM1 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PDLIM1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR