Anti-PDK4 Mouse Monoclonal Antibody [clone: 1A9]

H00005166-M02
103332-564EA 583.08 USD
103332-564
Anti-PDK4 Mouse Monoclonal Antibody [clone: 1A9]
The PDK4 Antibody (1A9) from Novus Biologicals is a mouse monoclonal antibody to PDK4. This antibody reacts with human. The PDK4 Antibody (1A9) has been validated for the following applications: ELISA.

Type: Primary
Antigen: PDK4
Clonality: Monoclonal
Clone: 1A9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR