Anti-PDK4 Mouse Polyclonal Antibody

H00005166-A02
103332-560EA 436.92 USD
103332-560
Anti-PDK4 Mouse Polyclonal Antibody
The PDK4 Antibody from Novus Biologicals is a mouse polyclonal antibody to PDK4. This antibody reacts with human. The PDK4 Antibody has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PDK4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR