Anti-PDK3 Mouse Monoclonal Antibody [clone: 1G11]

Supplier: Novus Biologicals
H00005165-M03
103332-558EA 798.46 USD
103332-558
Anti-PDK3 Mouse Monoclonal Antibody [clone: 1G11]
Antibodies
The PDK3 Antibody (1G11) from Novus Biologicals is a mouse monoclonal antibody to PDK3. This antibody reacts with human. The PDK3 Antibody (1G11) has been validated for the following applications: Western Blot, ELISA.

Type: Primary
Antigen: PDK3
Clonality: Monoclonal
Clone: 1G11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR