Anti-PDK2 Mouse Monoclonal Antibody [clone: 2G1]

Supplier: Novus Biologicals
H00005164-M01
103332-548EA 798.46 USD
103332-548
Anti-PDK2 Mouse Monoclonal Antibody [clone: 2G1]
Antibodies
The PDK2 Antibody (2G1) from Novus Biologicals is a mouse monoclonal antibody to PDK2. This antibody reacts with human. The PDK2 Antibody (2G1) has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry-Paraffin, RNA Inhibition.

Type: Primary
Antigen: PDK2
Clonality: Monoclonal
Clone: 2G1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR