Anti-PDK-1 Mouse Monoclonal Antibody [clone: 3B7]

H00005163-M01
103332-542EA 583.08 USD
103332-542
Anti-PDK-1 Mouse Monoclonal Antibody [clone: 3B7]
The PDK-1 Antibody (3B7) from Novus Biologicals is a mouse monoclonal antibody to PDK-1. This antibody reacts with human. The PDK-1 Antibody (3B7) has been validated for the following applications: ELISA.

Type: Primary
Antigen: PDK-1
Clonality: Monoclonal
Clone: 3B7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b Kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR