SP Wilmad-LabGlass O-Rings, Viton®, SP Industries
Supplier: SP Industries
LG-1022-576 LG-1022-520 LG-1022-538 LG-1022-542 LG-1022-518 LG-1022-552 LG-1022-526 LG-1022-546 LG-1022-524 LG-1022-539 LG-1022-550 LG-1022-522 LG-1022-562 LG-1022-572 LG-1022-534 LG-1022-566 LG-1022-514 LG-1022-504 LG-1022-571 LG-1022-556 LG-1022-516 LG-1022-530 LG-1022-574 LG-1022-554 LG-1022-506 LG-1022-557 LG-1022-544 LG-1022-568 LG-1022-502 LG-1022-564 LG-1022-536 LG-1022-578 LG-1022-532 LG-1022-528 LG-1022-560 LG-1022-512 LG-1022-558 LG-1022-540
14211-664EA 0.63 USD
14211-664 14211-642 14201-412 14211-640 14211-662 14211-624 14211-666 14201-396 14201-394 14201-372 14211-626 14212-052 14212-076 14212-078 14210-208 14211-672 14201-506 14201-408 14201-406 14211-632 14201-404 14201-402 14211-658 14211-656 14211-634 14212-080 14212-060 14201-384 14211-618 14211-518 14212-040 14201-382 14212-042 14212-086 14212-064 14212-088 14212-044 14210-458
SP Wilmad-LabGlass O-Rings, Viton®, SP Industries
These Viton® O-rings come in a variety of sizes.
  • Viton® is a fluorocarbon with low gas permeability, making it a good selection for high vacuum applications
  • Excellent chemical resistance over a wide temperature range (–20 to 200 °C)
  • Resists aliphatic and aromatic compounds
  • Viton® is a copolymer of hexafluoropropylene and 1,1-difluoroethylene, and it offers good mechanical and electrical properties
Caution: Not recommended for use with ketones, esters, and ethers.